missing translation for 'onlineSavingsMsg'
Learn More

ERCC1 Mouse anti-Human, Clone: ERCC1/2683, Novus Biologicals™

Product Code. 18359791 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359791 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18359791 Supplier Novus Biologicals Supplier No. NBP30716220UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ERCC1 Monoclonal specifically detects ERCC1 in Human samples. It is validated for Western Blot, ELISA, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERCC1
Applications Western Blot, ELISA, Peptide Array
Classification Monoclonal
Clone ERCC1/2683
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Alias COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20
Host Species Mouse
Immunogen Recombinant fragment (around aa 191-281) of human ERCC1 protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 2067
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.