missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC1 Mouse anti-Human, Clone: ERCC1/2318, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | ERCC1 |
|---|---|
| Clone | ERCC1/2318 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, ELISA, Protein Array |
| Applications | Western Blot, ELISA, Peptide Array |
Description
ERCC1 Monoclonal specifically detects ERCC1 in Human samples. It is validated for Western Blot, ELISA, Protein Array.Specifications
| ERCC1 | |
| 0.2 mg/mL | |
| Western Blot, ELISA, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 10 mM PBS with 0.05% BSA | |
| 2067 | |
| IgG2b κ | |
| Protein A or G purified |
| ERCC1/2318 | |
| Western Blot 1 to 2 μg/mL, ELISA, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20 | |
| Recombinant fragment (around aa 191-281) of human ERCC1 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title