missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC1 Antibody (ERCC1 3H11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £360.00
Specifications
| Antigen | ERCC1 |
|---|---|
| Clone | ERCC1 3H11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18576614
|
Novus Biologicals
NB100-117-0.025mg |
0.025 mg |
£150.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18555160
|
Novus Biologicals
NB100-117 |
0.1 mg |
£360.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ERCC1 Monoclonal antibody specifically detects ERCC1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| ERCC1 | |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| PBS | |
| 2067 | |
| IgG2a | |
| Protein A purified |
| ERCC1 3H11 | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20 | |
| Full length HIS-tagged recombinant human ERCC1 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title