missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC1 Antibody (8F1), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-704B
This item is not returnable.
View return policy
Description
ERCC1 Monoclonal specifically detects ERCC1 in Human, Rat, Chinese Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ERCC1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| ERCC1 | |
| Full length recombinant human ERCC1. [UniProt# P07992] | |
| 0.1 mL | |
| Primary | |
| Human, Rat, Chinese Hamster | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 8F1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20 | |
| Mouse | |
| Protein G purified | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 2067 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction