missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC1 Antibody (8F1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80725
This item is not returnable.
View return policy
Description
ERCC1 Monoclonal specifically detects ERCC1 in Human, Rat, Chinese Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ERCC1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000. Use reported in scientific literature, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (PMID 21961533), Immunohistochemistry-Paraffin | |
| COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, rat. Chinese Hamster reactivity reported in scientific literature (PMID: 17962301) Not yet tested in other species. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 8F1 | |
| Unconjugated | |
| PBS | |
| ERCC1 | |
| Full length recombinant human ERCC1. [UniProt# P07992] | |
| 0.1 mL | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 2067 | |
| Human, Rat, Chinese Hamster | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction