missing translation for 'onlineSavingsMsg'
Learn More

Ercalcidiol, MedChemExpress

Product Code. 16491919
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491919 1 mg 1mg
16470498 5 mg 5mg
16458038 10 mg 10mg
16453746 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491919 Supplier Medchem Express Supplier No. HY32349
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ercalcidiol is a metabolite of vitamin D2, is regarded as an indicator of vitamin D nutritional status.

Ercalcidiol is a metabolite of vitamin D2, is regarded as an indicator of vitamin D nutritional status.

Specifications

Chemical Name or Material Ercalcidiol
CAS 21343-40-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C28H44O2
Quantity 1 mg
Synonym 25-hydroxy Vitamin D2
Solubility Information DMSO : ≥ 100 mg/mL (242.34 mM)
SMILES C=C1CC[C@H](O)C/C1=C/C=C2[C@]3([C@@](C)([C@H](CC3)[C@@H](/C=C/[C@@H](C(C)(O)C)C)C)CCC/2)[H]
Molecular Weight (g/mol) 412.65
Formula Weight 412.65
Percent Purity 98.45%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.