missing translation for 'onlineSavingsMsg'
Learn More

ERC1 Mouse anti-Human, Mouse, Rat, Clone: 2A5B9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66681-1-IG-150UL

 View more versions of this product

Product Code. 16878785

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRAF-4 is a signal transducer associated with the cytoplasmic domain of the 75 kDa tumor necrosis factor receptor (TNF-R2).
Specifications

Specifications

ERC1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q811U3, Q8IUD2, Q99MI1
Erc1
ERC1 Fusion Protein Ag17665
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Western Blot
2A5B9
Unconjugated
ERC1
Cast2, ELKS, ERC 1, ERC1, KIAA1081, Rab6 interacting protein 2, RAB6IP2
Mouse
Protein G
RUO
111173, 23085, 266806
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.