missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERC1 Mouse anti-Human, Mouse, Rat, Clone: 2A5B9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66681-1-IG-20UL
This item is not returnable.
View return policy
Description
TRAF-4 is a signal transducer associated with the cytoplasmic domain of the 75 kDa tumor necrosis factor receptor (TNF-R2).Specifications
| ERC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q811U3, Q8IUD2, Q99MI1 | |
| Erc1 | |
| ERC1 Fusion Protein Ag17665 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 2A5B9 | |
| Unconjugated | |
| ERC1 | |
| Cast2, ELKS, ERC 1, ERC1, KIAA1081, Rab6 interacting protein 2, RAB6IP2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 111173, 23085, 266806 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction