missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) sulfate octahydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7050961
Click to view available options
Quantity:
5 g
10 g
50 g
This item is not returnable. View return policy
10031-52-4
MFCD00149696
erbium sulfate octahydrate, dierbium trisulfate octahydrate, erbium sulfate er2 so4 3 octahydrate, erbium sulfate hydrate er2 so4 3.8h2o, sulfuric acid, erbium 3+ salt 3:2 , octahydrate, erbium sulfate, erbium sulfate-water 2/3/8, erbium iii sulfate octahydrate reo, dierbium 3+ ion octahydrate trisulfate
This item is not returnable. View return policy
10031-52-4
MFCD00149696
erbium sulfate octahydrate, dierbium trisulfate octahydrate, erbium sulfate er2 so4 3 octahydrate, erbium sulfate hydrate er2 so4 3.8h2o, sulfuric acid, erbium 3+ salt 3:2 , octahydrate, erbium sulfate, erbium sulfate-water 2/3/8, erbium iii sulfate octahydrate reo, dierbium 3+ ion octahydrate trisulfate

Erbium(III) sulfate octahydrate is used as a colorant in glass manufacturing and porcelain enamel glazes. High purity Erbium Sulfate is applied as dopant in making optical fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Physical Form Crystalline
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 766.82 (622.70 Anhydrous)
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 3.205 g/mL
Assay (REO)
Chemical Name or Material Erbium(III) sulfate octahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'