missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) phosphate hydrate, 99.9% (REO), Thermo Scientific™

Product Code. 11344997
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11344997

Brand: Thermo Scientific Alfa Aesar 041993.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14242-01-4
Molecular Formula ErO4P
Molecular Weight (g/mol) 262.23
MDL Number MFCD00215989
InChI Key XFULIUKARWFBDF-UHFFFAOYSA-K
Synonym erbium iii phosphate hydrate, erbium iii phosphatehydrate, erbium 3+ hydrate phosphate, erbium iii phosphate hydrate trace metals basis
PubChem CID 16217375
SMILES [Er+3].[O-]P([O-])([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 10 g
IUPAC Name erbium(3+) phosphate
Formula Weight 262.23 (Anhydrous)
Percent Purity 99.9%
Odor Odorless
Assay (REO)
Chemical Name or Material Erbium(III) phosphate hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.