missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade

Product Code. 11490301
Change view
Click to view available options
Quantity:
∼2 g
∼50 g
10 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490301 10 g 10g
11480301 ∼2 g 2g
11400311 ∼50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 11490301 Supplier Thermo Scientific Alfa Aesar Supplier No. 040576.NC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

CAS: 14692-15-0 | Cl3ErH12O18 | 573.687 g/mol

Erbium(III) perchlorate hexahydrate is used in nuclear technology for neutron-absorbing control rods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14692-15-0
Molecular Formula Cl3ErH12O18
Molecular Weight (g/mol) 573.687
MDL Number MFCD00150232
InChI Key JZRNHNTYRRYOCR-UHFFFAOYSA-K
Synonym erbium iii perchlorate hexahydrate, erbium perchlorate-water 1/3/6, erbium iii perchlorate hexahydrate w/w in aqueous solution, reagent grade
PubChem CID 20038200
IUPAC Name erbium(3+);triperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Er+3]

Specifications

Color Pink
Physical Form Liquid
Quantity 10 g
UN Number UN3211
Solubility Information Micible with water.
Formula Weight 573.70 (465.61 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w in aq. soln.
Odor Odorless
Chemical Name or Material Erbium(III) perchlorate hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.