missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) perchlorate, 40 wt.% solution in water

Product Code. p-4089802 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10841671

Brand: Thermo Scientific Acros 428290050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14017-55-1 | Cl3ErO12 | 465.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Erbium(III) perchlorate
CAS 14017-55-1, 7732-18-5
Quantity 5 g
Molecular Formula Cl3ErO12
MDL Number MFCD00016073
Synonym erbium 3+ perchlorate, erbium perchlorate, perchloric acid,erbium 3+ salt 8ci,9ci, erbium iii perchlorate solution, perchloric acid erbium iii salt, erbium perchlorate aqueous solution
InChI Key FGBKFKCLWFRKEW-UHFFFAOYSA-K
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Er+3]
IUPAC Name erbium(3+);triperchlorate
Molecular Weight (g/mol) 465.61
PubChem CID 15335705
Formula Weight 465.61
Packaging Glass bottle
Specific Gravity 1.44
Density 1.4400g/mL
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Erbium(III) perchlorate, 40 wt.% solution in water
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.