missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) oxalate decahydrate, REacton™, 99.99% (REO), Thermo Scientific™

Product Code. 11386668
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11386668

Brand: Thermo Scientific Alfa Aesar 013216.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30618-31-6
Molecular Formula C6H26Er2O22
Molecular Weight (g/mol) 784.77
MDL Number MFCD00150142,MFCD00150142
InChI Key CYHKKMLSGVLNPD-UHFFFAOYSA-N
Synonym erbium oxalate decahydrate,
PubChem CID 131877482
IUPAC Name erbium;oxalic acid;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[Er].[Er].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O

Specifications

Physical Form Powder
Quantity 50 g
Linear Formula Er2(C2O4)3·10H2O
UN Number UN3288
Formula Weight 778.73 (598.58 Anhydrous)
Percent Purity 99.99%
Grade Reacton
Sensitivity Hygroscopic
Density 2.64 g/mL
Assay (REO)
Chemical Name or Material Erbium(III) oxalate decahydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.