missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) nitrate pentahydrate, REacton™, 99.99% (REO)

Product Code. 11381049
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381049 100 g 100g
11391049 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381049 Supplier Thermo Scientific Alfa Aesar Supplier No. 011306.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10031-51-3 | ErH10N3O14 | 443.35 g/mol

Erbium(III) nitrate pentahydrate is used as a chemical intermediates, producing a catalyst and related compounds, and also primary and secondary intermediated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-51-3
Molecular Formula ErH10N3O14
Molecular Weight (g/mol) 443.35
MDL Number MFCD00149691
InChI Key LWHHUEHWVBVASY-UHFFFAOYSA-N
Synonym erbium iii nitrate pentahydrate, erbium trinitrate pentahydrate, erbium nitrate pentahydrate, acmc-20al2v, er.3no3.5h2o, erbium nitrate-water 1/3/5, erbium iii nitrate pentahydrate, reacton, erbium nitrate, pentahydrate,
PubChem CID 53249207
SMILES O.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 130°C (in -4H2O)
Physical Form Crystalline Aggregates
Quantity 100 g
UN Number UN1477
Solubility Information Soluble in water.
IUPAC Name erbium(3+);trinitrate;pentahydrate
Formula Weight 443.35 (353.27 Anhydrous)
Percent Purity 99.99%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Erbium(III) nitrate pentahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.