missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) fluoride, anhydrous, REacton™, 99.99% (REO)

Product Code. 11361059
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361059 50 g 50g
11371059 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361059 Supplier Thermo Scientific Alfa Aesar Supplier No. 011312.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13760-83-3 | ErF3 | 224.25 g/mol

Erbium(III) fluoride is an important colorant in glass manufacturing and porcelain enamel glazes. It is used to produce erbium metal. It is used as dopant in making optical products. It is highly employed as erbium doped fluoride fibers in optical communications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13760-83-3
Molecular Formula ErF3
Molecular Weight (g/mol) 224.25
MDL Number MFCD00016074
InChI Key QGJSAGBHFTXOTM-UHFFFAOYSA-K
IUPAC Name erbium(3+) trifluoride
SMILES [F-].[F-].[F-].[Er+3]

Specifications

Melting Point 1350°C
Physical Form Granules, Powder
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 224.28
Percent Purity 99.99%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Density 6.4 g/mL
Assay (REO)
Chemical Name or Material Erbium(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.