missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) chloride hydrate, REacton™, 99.9% (REO)

Product Code. 11341049
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341049 100 g 100g
11351049 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341049 Supplier Thermo Scientific Alfa Aesar Supplier No. 011304.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19423-85-9 | Cl3Er | 273.61 g/mol

Erbium(III) chloride hydrate is used as a starting material for new edge-bridged octahedral M6 cluster compounds such as CsErTa6Cl18, for which electron and stability studies were performed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19423-85-9
Molecular Formula Cl3Er
Molecular Weight (g/mol) 273.61
MDL Number MFCD00010989
InChI Key HDGGAKOVUDZYES-UHFFFAOYSA-K
IUPAC Name erbium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Er+3]

Specifications

Physical Form Crystalline Aggregates
Quantity 100 g
Solubility Information Slightly soluble in ethanol
Formula Weight 273.62 (Anhydrous)
Percent Purity 99.9%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Erbium(III) chloride hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.