Learn More
Erbium(III) chloride, anhydrous, 99.9% (metals basis)
Description
It is a starting material for new edge-bridged octahedral M6 cluster compounds which are used for electron and stability studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1500°C |
| Melting Point | 774°C |
| Physical Form | Beads |
| Quantity | 250 g |
| Solubility Information | Soluble in water,strong mineral acids. Slightly soluble in MeOH and THF. |
| Formula Weight | 273.62 |
| Percent Purity | 99.9% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Assay | (metals basis) |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.