missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erbium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals
216.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
250 g
Description
It is a starting material for new edge-bridged octahedral M6 cluster compounds which are used for electron and stability studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1500°C |
Melting Point | 774°C |
Physical Form | Beads |
Quantity | 50 g |
Solubility Information | Soluble in water,strong mineral acids. Slightly soluble in MeOH and THF. |
Formula Weight | 273.62 |
Percent Purity | 99.9% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Assay | (metals basis) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'