missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) acetate tetrahydrate, REacton™, 99.9% (REO)

Product Code. 11378458
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378458 5 g 5g
11388458 25 g 25g
11368458 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378458 Supplier Thermo Scientific Alfa Aesar Supplier No. 040481.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 15280-57-6

Erbium(III) acetate tetrahydrate participates in the synthesis and determination of crystal structures and stereochemical properties of lanthanide(III) complexes with ethylenediamine-N,N,N?,N?-tetraacetate. Participates in simple one-pot single-step synthesis of rare earth doped spherical mesoporous metal oxide nanospheres with upconversion emission ability in supercritical methanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15280-57-6
MDL Number MFCD00013034

Specifications

Physical Form Crystalline Aggregates
Quantity 5 g
Linear Formula Er(OOCCH3)3·4H2O
Solubility Information Soluble in water.
Formula Weight 416.45 (344.39 Anhydrous)
Percent Purity 99.9%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Density 2.114 g/mL
Assay (REO)
Chemical Name or Material Erbium(III) acetate tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.