missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Erbin Polyclonal Antibody
GREENER_CHOICE

Sheep Polyclonal Antibody

Brand:  Invitrogen™ PA5149414

Product Code. 30533647

  • £415.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Erbin
Polyclonal
Unconjugated
ERBIN
1700028E05Rik; densin-180-like protein; epididymis secretory protein Li 78; Erbb2 interacting protein; ERBB2 interacting protein-like; erbb2-interacting protein; Erbb2ip; Erbin; HEL-S-78; Kiaa1225; Lap2; mKIAA1225; Protein LAP2; RGD1562952; similar to Erbb2 interacting protein isoform 2
Sheep
Antigen affinity chromatography
RUO
55914
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with trehalose and no preservative
Q96RT1
ERBIN
E. coli-derived recombinant human Erbin, Lys496-Thr687.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.