missing translation for 'onlineSavingsMsg'
Learn More

Erbin Antibody (10G1), Novus Biologicals™

Product Code. 18349309 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349309 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349309 Supplier Novus Biologicals Supplier No. H00055914M01100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Erbin Monoclonal antibody specifically detects Erbin in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Erbin
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 10G1
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_061165
Gene Alias Densin-180-like protein, erbb2 interacting protein, Erbb2-interacting proteinErbin, ERBINLAP2protein LAP2, KIAA1225
Host Species Mouse
Immunogen ERBB2IP (NP_061165, 1272 a.a. ~ 1371 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. QGHELAKQEIRVRVEKDPELGFSISGGVGGRGNPFRPDDDGIFVTRVQPEGPASKLLQPGDKIIQANGYSFINIEHGQAVSLLKTFQNTVELIIVREVSS
Purification Method IgG purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55914
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.