missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB4 Monoclonal Antibody (H4.77.16 (Ab77)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512888
This item is not returnable.
View return policy
Description
MA5-12888 targets HER-4 in Western blot, flow cytometry and immunoprecipitation applications and shows reactivity with Human, mouse, and Rat samples. The MA5-12888 immunogen is extracellular fragment of recombinant human c-erbB-4/HER-4 oncoprotein.
Oncogene ERBB4 (HER4) is a member of the type I receptor protein tyrosine subfamily that includes EGFR, ERBB2, and ERBB3. ERBB4 binds and is activated by neuregulins and other factors and induces a variety of cellular responses including mitogenesis and differentiation. ErbB 4 is activated by neuregulins and NTAK (neural and thymus derived activator for ErbB kinases). ErbB 4 is a receptor for heregulin and is capable of mediating HGL-stimulated tyrosine phosphorylation of itself. Like other protein kinases, ErbB 4 mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. ErbB 4 is most predominantly expressed in several breast carcinoma cell lines, and in normal skeletal muscle, heart, pituitary, brain, and cerebellum. Breast tumor cell lines T47-D, MDA-MB-453, BT-474 and H3396 are found to have the highest levels of mRNA, and intermediate levels are seen in MCF-7, MDAMB-330 and MDA-MB-361. Expression of ErbB 4 is low or absent in some breast tumor cell lines such as MDA-MB-231, MDA-MB-157, MDA-MB-468, and SKBR-3. ErbB 4 levels have been found to be elevated in certain human tumor cell lines suggesting that it may play a role in some malignancies.
Specifications
| ErbB4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| Q15303, Q61527, Q62956 | |
| ERBB4 | |
| Extracellular fragment of recombinant human c-erbB-4/HER-4 oncoprotein | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunoprecipitation | |
| H4.77.16 (Ab77) | |
| Unconjugated | |
| ERBB4 | |
| 4ICD; ALS19; avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4; avian erythroblastosis oncogene B 4; c-erbB-4; erb-b2 receptor tyrosine kinase 4; Erbb4; ERBB4 intracellular domain; ERBB4 transcript variant I12DEL; ERBB4 transcript variant I20DEL; Her4; human epidermal growth factor receptor 4; Mer4; OTTHUMP00000209432; p180erbB4; proto-oncogene-like protein c-ErbB-4; receptor tyrosine kinase; receptor tyrosine-protein kinase erbB-4; s80HER4; Tyro-2; Tyrosine kinase-type cell surface receptor HER4; v-erb-a erythroblastic leukemia viral oncogene homolog 4; v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian); v-erb-b2 avian erythroblastic leukemia viral oncogene 4; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 4 | |
| Mouse | |
| Protein G | |
| RUO | |
| 13869, 2066, 59323 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction