missing translation for 'onlineSavingsMsg'
Learn More

ErbB4/Her4 Rabbit anti-Mouse, Rat, Clone: 8E5D0, Novus Biologicals™

Product Code. 18354657
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354657 100 μg 100µL
18353593 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18354657 Supplier Novus Biologicals Supplier No. NBP315632100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ErbB4/Her4 Monoclonal antibody specifically detects ErbB4/Her4 in Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ErbB4/Her4
Applications Western Blot
Classification Monoclonal
Clone 8E5D0
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4, EC 2.7.10, EC 2.7.10.1, HER4MGC138404, p180erbB4, Proto-oncogene-like protein c-ErbB-4, receptor tyrosine-protein kinase erbB-4, Tyrosine kinase-type cell surface receptor HER4, v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 4, v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1200-1308 of human ErbB4/Her4 (Q15303).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Protein Kinase, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2066
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.