missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB4/Her4 Mouse anti-Human, Mouse, Clone: HFR-1, Novus Biologicals™
Mouse Monoclonal Antibody
£405.00
Specifications
| Antigen | ErbB4/Her4 |
|---|---|
| Clone | HFR-1 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL |
| Applications | Flow Cytometry, Immunofluorescence |
Description
ErbB4/Her4 Monoclonal specifically detects ErbB4/Her4 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
| ErbB4/Her4 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors | |
| 10 mM PBS with 0.05% BSA | |
| 2066 | |
| IgG2b κ | |
| Protein A or G purified |
| HFR-1 | |
| Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4, EC 2.7.10, EC 2.7.10.1, HER4MGC138404, p180erbB4, Proto-oncogene-like protein c-ErbB-4, receptor tyrosine-protein kinase erbB-4, Tyrosine kinase-type cell surface receptor HER4, v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 4, v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian) | |
| Recombinant human ErbB4/Her4 protein fragment (around aa 1116-1269) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title