missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB4/Her4 Antibody (JE59-43), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32314
This item is not returnable.
View return policy
Description
ErbB4/Her4 Monoclonal antibody specifically detects ErbB4/Her4 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| ErbB4/Her4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4, EC 2.7.10, EC 2.7.10.1, HER4MGC138404, p180erbB4, Proto-oncogene-like protein c-ErbB-4, receptor tyrosine-protein kinase erbB-4, Tyrosine kinase-type cell surface receptor HER4, v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 4, v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian) | |
| Synthetic peptide within Human ErbB4/Her4 aa 1,231-1,280/ 1,308. (Uniprot: Q15303) | |
| 100 μL | |
| Angiogenesis, Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors | |
| 2066 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE59-43 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction