Learn More
Invitrogen™ ErbB4 (HER4)
Oncogene ERBB4 (HER4) is a member of the type I receptor protein tyrosine subfamily that includes EGFR, ERBB2, and ERBB3.
Brand: Invitrogen™ PR6308B
Code : NEW
Additional Details : Weight : 0.01000kg
Description
ERBB4 binds and is activated by neuregulins and other factors and induces a variety of cellular responses including mitogenesis and differentiation.
Specifications
NP_005226 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
ErbB4 (HER4) | |
Human | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
61.6kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
EGF Receptor (ErbB) Kinase Family | |
GST Tag | |
Frozen |