Learn More
ERBB3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30452-1-AP-150UL
This item is not returnable.
View return policy
Description
The ERBB3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ERBB3. This antibody recognizes human antigen. The ERBB3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ERBB3 | |
| 350 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| c erbB 3, c erbB3, ErbB 3, ERBB3, erbB3 S, HER3, LCCS2, MDA BF 1, p180 ErbB3, p45 sErbB3, p85 sErbB3 | |
| 150 μL | |
| 2065 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |