missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB3 Monoclonal Antibody (H3.105.5 (Ab105)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513008
This item is not returnable.
View return policy
Description
MA5-13008 targets HER-3 in WB, flow cytometry, ICC/IF, and IP applications and shows reactivity with Human and mouse samples. The MA5-13008 immunogen is extracellular domain of recombinant human c-erbB-3/HER-3 oncoprotein.
ErbB3 Monoclonal antibody specifically detects ErbB3 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot.
Specifications
| ErbB3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P21860, Q61526 | |
| ERBB3 | |
| Extracellular domain of recombinant human c-erbB-3/HER-3 oncoprotein | |
| 500 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| H3.105.5 (Ab105) | |
| Unconjugated | |
| ERBB3 | |
| avian erythroblastosis oncogene B 3; avian erythroblastosis oncogene B 3 receptor; C76256; c-erbB3; c-erbB-3; C-errB-3; erb-b2 receptor tyrosine kinase 3; ERBB3; Erbb-3; Erbb3r; erbB3-S; glial growth factor receptor; HER3; human epidermal growth factor receptor 3; LCCS2; MDA BF 1; MDA-BF-1; MGC88033; nuc-ErbB3; p180 ErbB3; p180-ErbB3; p45 sErbB3; p45-sErbB3; p85 sErbB3; p85-sErbB3; proto-oncogene-like protein c-ErbB-3; receptor tyrosine kinase; receptor tyrosine-protein kinase erbB-3; receptor tyrosine-protein kinase erbB-3; LOW QUALITY PROTEIN: receptor tyrosine-protein kinase erbB-3; tyrosine kinase-type cell surface receptor HER3; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) | |
| Mouse | |
| Protein G | |
| RUO | |
| 13867, 2065 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction