Learn More
ErbB3/Her3 Antibody (S05-3F7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19862-50ul
 View more versions of this product
Product Code. 18628873
  • £328.00 / 50µL
Description
ErbB3/Her3 Monoclonal antibody specifically detects ErbB3/Her3 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ErbB3/Her3
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse
Purified
Western Blot
S05-3F7
Unconjugated
c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
A synthetic peptide of human ErbB3/Her3 (Uniprot # P21860)
50 μg
Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors
2065
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.