missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB3 (HER-3) Mouse anti-Human, BV750, Clone: SGP1, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 751783
This item is not returnable.
View return policy
Description
ErbB3 (HER-3) Monoclonal antibody specifically detects ErbB3 (HER-3) in Human samples. It is validated for Flow Cytometry.
Specifications
| ErbB3 (HER-3) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Hu ErbB3 (HER-3) BV750 SGP1 50 μg | |
| Human cEerbB3 Recombinant Protein | |
| RUO | |
| ErbB3 (HER-3) | |
| Refrigerate (2 to 8°C) | |
| Human |
| Flow Cytometry | |
| SGP1 | |
| Brilliant Violet 750 | |
| Mouse | |
| 1 x 50 μg | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction