missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB3 (HER-3) Mouse anti-Human , Alexa Fluor 647, Clone: SGP1, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 567044
This item is not returnable.
View return policy
Description
ErbB3 (HER-3) Monoclonal antibody specifically detects ErbB3 (HER-3) in Human samples. It is validated for Flow Cytometry
Specifications
| ErbB3 (HER-3) | |
| Monoclonal | |
| Alexa Fluor 647 | |
| ERBB3; ErbB-3; HER3; c-erbB-3; c-erbB3; human epidermal growth factor receptor 3; receptor tyrosine-protein kinase erbB-3 | |
| Human cEerbB3 Recombinant Protein | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| SGP1 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 647 under optimum conditions, and unreacted Alexa Fluor 647 was removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction