missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX ErbB2 Protein, Active

Product Code. 17340824
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17340824

Brand: Eurofins DiscoverX 14939

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-terminal 6Jis-tagged, recombinant, human ErbB2, for use in Kinase Assays.

Human EGF receptor 2 (HER2, ERBB2) tyrosine kinase is an EGF receptor-related protein tyrosine kinase which is overexpressed in many mammary and ovarian carcinomas. Binding of the ligand to the receptor is believed to activate the receptor's intrinsic protein kinase activity, initiating a signal transduction pathway which leads to increased cell proliferation. Truncated HER2, lacking the extracellular and transmembrane regions, is constitutively active. The active kinase transfers g-phosphates of ATP to the phenolic hydroxyl of specific tyrosine residues on target proteins.

GenBank NM_004448

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.01% CHAPS, 50% glycerol, 2 mM DTT. Frozen solution.
Molecular Weight (g/mol) 69 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag C-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Truncated
Purity or Quality Grade ≥71%
Protein ErbB2, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.