missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB2/Her2 Rabbit anti-Human, Clone: 7N1S0, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £432.00
Specifications
| Antigen | ErbB2/Her2 |
|---|---|
| Clone | 7N1S0 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18326412
|
Bio-Techne
NBP3-16960-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18324526
|
Novus Biologicals
NBP3-16960-100UL |
100 μg |
£432.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ErbB2/Her2 Monoclonal antibody specifically detects ErbB2/Her2 in Human samples. It is validated for Western BlotSpecifications
| ErbB2/Her2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Cellular Markers, Core ESC Like Genes, Oncogenes, Phospho Specific, Protein Kinase, Stem Cell Markers, Tumor Suppressors | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 2064 | |
| IgG | |
| Affinity purified |
| 7N1S0 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian) | |
| A synthetic phosphorylated peptide around Y1139 of human ErbB2/Her2 (P04626). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title