missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB2 (HER-2), His Tag ErbB2 (HER2), His Tag Recombinant Protein
ErbB2 (HER-2), His Tag ErbB2 (HER2), His Tag Recombinant Protein
£439.00 - £6890.00
Specifications
Accession Number | NP_004439 |
---|---|
Gene Alias | ErbB2 (HER2), NEU |
Species | Human |
Protein Tag | His Tag |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11523447
|
Invitrogen
PV3366 |
10μg |
£439.00
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15478423
|
Invitrogen
PR4918A |
100μg |
£1602.00
100µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10083182
|
Invitrogen
PV3590 |
1mg |
£6890.00
1mg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ErbB2 (HER2) is a receptor tyrosine kinase that is overexpressed in some breast tumors. Herceptin, used in treatment of metastatic Her2-positive cancer, is monoclonal antibody targeting this kinase.Specifications
NP_004439 | |
Human | |
Baculovirus |
ErbB2 (HER2), NEU | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.