missing translation for 'onlineSavingsMsg'
Learn More

Erastin, MedChemExpress

Product Code. 16489347
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489347 5 mg 5mg
16452877 50 mg 50mg
16436808 10 mg 10mg
16406926 100 mg 100mg
16471196 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16489347 Supplier Medchem Express Supplier No. HY15763
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Erastin is a ferroptosis inducer. Erastin binds and inhibits voltage-dependent anion channels (VDAC2/VDAC3).

Erastin is a ferroptosis inducer. Erastin binds and inhibits voltage-dependent anion channels (VDAC2/VDAC3).

Specifications

Chemical Name or Material Erastin
CAS 571203-78-6
Color White
Purity Grade Notes Research
Molecular Formula C30H31ClN4O4
Quantity 5 mg
Solubility Information DMSO : 12.5 mg/mL (22.85 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1N(C2=CC=CC=C2OCC)C(C(N3CCN(C(COC4=CC=C(Cl)C=C4)=O)CC3)C)=NC5=C1C=CC=C5
Molecular Weight (g/mol) 547.04
Formula Weight 547.04
Percent Purity 98.46%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.