missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERAS Monoclonal Antibody (D10-B9)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA532907
This item is not returnable.
View return policy
Description
ERAS Monoclonal Antibody for ICC/IF, IHC (P)
The human ERAS gene was initially reported as a processed pseudogene (HRasp, Ha-Ras2), with several base substitutions in coding regions, but was more recently described as potentially encoding a functional human ERAS protein. The ERAS oncogene supports the tumorigenic growth of embryonic stem cells by producing a constitutively active Ras protein. ERAS protein has less than 50% amino acid identity with H-Ras, another member of Ras family but the key domains essential for Ras function are highly conserved. The protein also contains amino-acid residues identical to those present in active mutants of Ras and causes oncogenic transformation in NIH 3T3 cells. Human ERAS gene is localized in the chromosomal region Xp11.23.
Specifications
| ERAS | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| Q7Z444 | |
| ERAS | |
| Recombinant protein within Human ERAS aa 1-233. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry | |
| D10-B9 | |
| Unconjugated | |
| ERAS | |
| ecat5; Embryonic stem cell-expressed Ras; ERAS; E-Ras; ES cell expressed Ras; ES cell-expressed Ras; GTPase ERas; Ha-Ras2; HRAS2; HRasp; small GTPase E-Ras; small GTPase protein E-Ras; v-Ha-ras Harvey rat sarcoma viral oncogene homolog 2; v-Ha-ras Harvey rat sarcoma viral oncogene homolog pseudogene | |
| Mouse | |
| Protein A | |
| RUO | |
| 3266 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction