missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERAS Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99426CL1
This item is not returnable.
View return policy
Description
ERAS Polyclonal antibody specifically detects ERAS in Mouse samples. It is validated for Western Blot
Specifications
| ERAS | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| Embryonic stem cell-expressed Ras, E-Ras, ES cell expressed Ras, HRAS2MGC126693, HRASPGTPase ERas, MGC126691, small GTPase protein E-Ras, v-Ha-ras Harvey rat sarcoma viral oncogene homolog 2, v-Ha-ras Harvey rat sarcoma viral oncogene homolog pseudogene | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ERAS | |
| 0.1 mL | |
| Signal Transduction | |
| 3266 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction