missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERAB Mouse anti-Human, Mouse, Clone: 1G5E5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60069-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes 3-hydroxyacyl-CoA dehydrogenase type II, a member of the short-chain dehydrogenase/reductase superfamily. The gene product is a mitochondrial protein that catalyzes the oxidation of a wide variety of fatty acids, alcohols, and steroids. The protein has been implicated in the development of Alzheimer′s disease, and mutations in the gene are the cause of 2-methyl-3-hydroxybutyryl-CoA dehydrogenase deficiency (MHBD). Several alternatively spliced transcript variants have been identified, but the full-length nature of only two transcript variants has been determined.Specifications
| ERAB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O08756, Q99714 | |
| Hsd17b10 | |
| ERAB Fusion Protein Ag1020 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 1G5E5 | |
| Unconjugated | |
| HSD17B10 | |
| 17 beta HSD 10, 17b HSD10, ABAD, CAMR, DUPXp11.22, ERAB, HADH2, HCD2, HSD17B10, MHBD, MRPP2, MRX17, MRX31, MRXS10, SCHAD, SDR5C1, Type II HADH, XH98G2 | |
| Mouse | |
| Caprylic Acid Precipitation | |
| RUO | |
| 15108, 3028 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction