missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERAB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38154-100ul
This item is not returnable.
View return policy
Description
ERAB Polyclonal antibody specifically detects ERAB in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| ERAB | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 1:50 - 1:100 | |
| 17-beta-HSD 10, 17-beta-hydroxysteroid dehydrogenase 10, 17b-HSD10, 3-hydroxy-2-methylbutyryl-CoA dehydrogenase, ABAD, amyloid-beta peptide binding alcohol dehydrogenase, CAMR, DUPXp11.22, EC 1.1.1.178, EC 1.1.1.35, Endoplasmic reticulum-associated amyloid beta-peptide-binding protein, ERAB3-hydroxyacyl-CoA dehydrogenase type-2, HADH2AB-binding alcohol dehydrogenase, hydroxyacyl-Coenzyme A dehydrogenase, type II, hydroxyacyl-Coenzyme Adehydrogenase, type II, hydroxysteroid (17-beta) dehydrogenase 10, mental retardation, X-linked, syndromic 10, MHBD, Mitochondrial ribonuclease P protein 2,3-hydroxyacyl-CoA dehydrogenase type II, Mitochondrial RNase P protein 2, MRPP2HCD2, MRX17, MRX31, MRXS10, SCHAD, SDR5C1, short chain dehydrogenase/reductase family 5C, member 1, short chain L-3-hydroxyacyl-CoA dehydrogenase type 2, short chain type dehydrogenase/reductase XH98G2, Short-chain type dehydrogenase/reductase XH98G2, Type II HADH, XH98G2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-261 of human ERAB (NP_004484.1).,, Sequence:, MAAACRSVKGLVAVITGGASGLGLATAERLVGQGASAVLLDLPNSGGEAQAKKLGNNCVFAPADVTSEKDVQTALALAKGKFGRVDVAVNCAGIAVASKTYNLKKGQTHTLEDFQRVLDVNLMGTFNVIRLVAGEMGQNEPDQGGQRGVIINTASVAAFEGQVGQAAYSASKGGIVGMTLPIARDLAPIGIRVMTIAPGLFGTPLLTSLPEKVCNFLASQVPFPSRLGDPAEYAHLVQAIIENPFLNGEVIRLDGAIRMQP | |
| 100 μL | |
| Cancer, Lipid and Metabolism | |
| 3028 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction