missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERAB Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NB100-75460
This item is not returnable.
View return policy
Description
ERAB Polyclonal specifically detects ERAB in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| ERAB | |
| Polyclonal | |
| Western Blot | |
| Q16836 | |
| HSD17B10 | |
| The immunogen for this antibody was a sequence from 57a.a. to 314a.a. from ERAB(a 314a.a. long protein) | |
| 0.05 mg | |
| Lipid and Metabolism | |
| 3028 | |
| Human, Mouse, Rat |
| Western Blot | |
| Unconjugated | |
| PBS with No Preservative | |
| 17-beta-HSD 10, 17-beta-hydroxysteroid dehydrogenase 10, 17b-HSD10, 3-hydroxy-2-methylbutyryl-CoA dehydrogenase, ABAD, amyloid-beta peptide binding alcohol dehydrogenase, CAMR, DUPXp11.22, EC 1.1.1.178, EC 1.1.1.35, Endoplasmic reticulum-associated amyloid beta-peptide-binding protein, ERAB3-hydroxyacyl-CoA dehydrogenase type-2, HADH2AB-binding alcohol dehydrogenase, hydroxyacyl-Coenzyme A dehydrogenase, type II, hydroxyacyl-Coenzyme Adehydrogenase, type II, hydroxysteroid (17-beta) dehydrogenase 10, mental retardation, X-linked, syndromic 10, MHBD, Mitochondrial ribonuclease P protein 2,3-hydroxyacyl-CoA dehydrogenase type II, Mitochondrial RNase P protein 2, MRPP2HCD2, MRX17, MRX31, MRXS10, SCHAD, SDR5C1, short chain dehydrogenase/reductase family 5C, member 1, short chain L-3-hydroxyacyl-CoA dehydrogenase type 2, short chain type dehydrogenase/reductase XH98G2, Short-chain type dehydrogenase/reductase XH98G2, Type II HADH, XH98G2 | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| Primary | |
| ERAB | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction