missing translation for 'onlineSavingsMsg'
Learn More

ER Stress/ UPR Antibody Pack, Novus Biologicals™

Product Code. 18653959 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
8 Vials
Unit Size:
Pack of 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18653959 8 Vials Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18653959 Supplier Novus Biologicals Supplier No. NBP252746

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody Pack

ER Stress / UPR antibody specifically detects ER Stress / UPR in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ER Stress / UPR
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Gene Alias anti-Growth arrest and DNA damage-inducible protein GADD153, ire1 alpha p Ser724, phospho ire1 alpha, phospho-ire1 alpha, p-ire1 alpha
Immunogen See individual datasheets.
Quantity 8 Vials
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cellular Markers, ER Markers, Signal Transduction, Unfolded Protein Response, Unfolded Protein Response (UPR)
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.