missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ER-beta1 Mouse, Clone: ERb455, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2100-MSM1-P1
This item is not returnable.
View return policy
Specifications
| ER-beta1 | |
| Predominantly nuclear | |
| ERb455 | |
| Unconjugated | |
| NP_001428 | |
| Mouse | |
| 53-59kDa | |
| Protein A/G Purified | |
| RUO | |
| 2100 | |
| IgG2a |
| Flow Cytometry, Immunofluorescence, Western Blot, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| ESR2 | |
| C-terminus fragment of recombinant human estrogen receptor beta protein | |
| MCF-7 Cells. Ovarian, Breast, Bladder, Gastric or Salivary carcinoma | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction