missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Estrogen Receptor beta (ER beta) ER Beta Recombinant Protein

Product Code. 15178156
Click to view available options
Quantity:
180 μg
Unit Size:
1.8µg
This item is not returnable. View return policy

Product Code. 15178156

Brand: Invitrogen™ A15664

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. ER beta full-length recombinant protein is: , Flexible—use in your in-house assay or as part of our PolarScreen™ ER Beta Competitive Binding Assay for a complete assay solution, Ready to use—active, purified, and native (untagged), Convenient—available in bulk sizes ER beta recombinant protein is manufactured under the strictest quality parameters. This product is guaranteed for 6 months after purchase. Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond. Protein Form: Recombinant, Full Length

Estrogen Receptor beta (ER beta, NR3A2) protein is approximately 55kD, though multiple isoforms ranging from 36-59kD have been described. Human ER beta was initially cloned and characterized from testis. Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Estrogen receptor beta (ER beta) binds estrogens with an affinity similar to that of estrogen receptor alpha. Some ER beta isoforms dominantly inhibit the activity of estrogen receptor alpha in reproductive tissues. ER beta is found widely in many tissues throughout the body and can act as a potent tumor suppressor, playing a crucial role in many cancer types such as prostate cancer, autism spectrum disorder, medullary thyroid carcinoma, De Quervain Disease and risk of cardiovascular disease.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92731
Concentration See Label
For Use With (Application) Functional Assay
Formulation proprietary buffer with no preservative
Gene ID (Entrez) 2100
Molecular Weight (g/mol) 53.4 kDa
Name Estrogen Receptor beta (ER beta) ER Beta
Quantity 180 μg
Regulatory Status RUO - research use only
Gene Alias ER; ER beta; ER[b]; Erb; Erb2; Erbeta; ER-beta; Esr2; ESRB; ESR-BETA; ESTRB; estrogen receptor 2; estrogen receptor 2 (beta); estrogen receptor 2 (ER beta); estrogen receptor 2 beta; estrogen receptor beta; estrogen receptor beta 1 isoform deletion variant; estrogen receptor beta 4; NR3A2; Nuclear receptor subfamily 3 group A member 2; oestrogen receptor beta; steroid receptor
Common Name Estrogen Receptor beta
Protein Length 477
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.