missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ER beta/NR3A2 Antibody (57/3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£160.00 - £392.00
Specifications
| Antigen | ER beta/NR3A2 |
|---|---|
| Clone | 57/3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18290514
|
Novus Biologicals
NBP2-61943 |
0.1 mg |
£392.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663567
|
Novus Biologicals
NBP2-61943-0.025mg |
0.025 mg |
£160.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ER beta/NR3A2 Monoclonal specifically detects ER beta/NR3A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| ER beta/NR3A2 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| PBS with 0.02% Sodium Azide | |
| 2100 | |
| Recognizes ER beta2 isomer at sequence CGMKMETLLPEATMEQ | |
| Primary | |
| 53 kDa |
| 57/3 | |
| Western Blot 1:500, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
| Erb, ER-beta, ESR-BETA, ESTRB, estrogen receptor 2 (ER beta), estrogen receptor beta, estrogen receptor beta 4, NR3A2ESRB, Nuclear receptor subfamily 3 group A member 2 | |
| ESR2 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title