missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ER alpha/NR3A1, p Ser106 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£213.00 - £350.00
Specifications
| Antigen | ER alpha/NR3A1 (p Ser106) |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18280372
|
Novus Biologicals
NB100-81912-0.025ML |
0.025 mL |
£213.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18299840
|
Novus Biologicals
NB100-81912 |
0.05 mL |
£350.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ER alpha/NR3A1 Polyclonal specifically detects ER alpha/NR3A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ER alpha/NR3A1 (p Ser106) | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| P03372 | |
| 2099 | |
| IgG | |
| Affinity Purified | |
| 66 kDa |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Rat, Human, Mouse | |
| ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
| ESR1 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title