missing translation for 'onlineSavingsMsg'
Learn More

ER alpha/NR3A1 Antibody (h-151), Novus Biologicals™

Product Code. 18211754 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
Unit Size:
0.1mg
25µg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18211754 0.025 mg 25µg
18230181 0.1 mg 0.1mg
18553924 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18211754 Supplier Novus Biologicals Supplier No. NB120135380.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ER alpha/NR3A1 Monoclonal specifically detects ER alpha/NR3A1 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ER alpha/NR3A1
Applications Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone h-151
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:250, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 15 ug/ml
Formulation PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
Gene Accession No. P03372
Gene Alias ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1
Gene Symbols ESR1
Host Species Mouse
Immunogen Synthetic peptide derived from the sequence of human Estrogen Receptor, conjugated to KLH
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2099
Test Specificity Binding specificity was confirmed in peptide inhibition studies with the intact protein. This does not recognize the native estrogen receptor in the inactive 8S oligomeric state, but reacts efficiently with the activated 4S receptor.
Target Species Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.