missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ER alpha/NR3A1 Antibody [FITC], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB200-347F
This item is not returnable.
View return policy
Description
ER alpha/NR3A1 Polyclonal antibody specifically detects ER alpha/NR3A1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| ER alpha/NR3A1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| FITC | |
| ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
| The immunogen recognized by this antibody maps to a region between residue 550 and the C-terminus (residue 595) of human Estrogen Receptor Alpha using the numbering given in SwissProt entry P03372 (GeneID 2099). | |
| 0.1 mL | |
| Angiogenesis, Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 2099 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction