missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ER alpha/NR3A1 Antibody (ESR1/8407R) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20981
This item is not returnable.
View return policy
Description
ER alpha/NR3A1 Monoclonal antibody specifically detects ER alpha/NR3A1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ER alpha/NR3A1 | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
| Recombinant fragment (around aa495-595) of human ER alpha/NR3A1 protein (exact sequence is proprietary) | |
| 100 μg | |
| Angiogenesis, Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 2099 | |
| Store at -20°C to -70°C. Avoid freeze/thaw cycles. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| ESR1/8407R | |
| Unconjugated | |
| 10 mM PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction