missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ER alpha/NR3A1 Antibody (ESR1/420), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-48011UV
This item is not returnable.
View return policy
Description
ER alpha/NR3A1 Monoclonal antibody specifically detects ER alpha/NR3A1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, CyTOF
Specifications
| ER alpha/NR3A1 | |
| Monoclonal | |
| DyLight 350 | |
| ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
| Recombinant human ER alpha/NR3A1 protein (aa290-560) (Uniprot: P03372) | |
| 0.1 mL | |
| Angiogenesis, Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
| 2099 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF | |
| ESR1/420 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction