missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ER 27319 maleate
Description
Selective inhibitor of Syk kinase. Inhibits tyrosine phosphorylation of Syk initiated by the engagement of FcεRI in rat and human mast cells. Results in the abrogation of degranulation, TNF-α production (IC50 = 10 μM) and other related signaling events.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 396.17 |
| Purity | >99% |
| Molecular Weight (g/mol) | 396.443 |
| Chemical Name or Material | ER 27319 maleate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?